Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
£0.00
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
United Kingdom
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
1,1,2,2-Tetrafluoro-3-methoxypropane
1,1,2,2-Tetrafluoro-3-methoxypropane
1,1,2,2-Tetrafluoro-3-methoxypropane
APOSPC10613-25G
:
Apollo Scientific
:
1,1,2,2-Tetrafluoro-3-methoxypropane
APOSPC10613-25G
:
Apollo Scientific
:
Pack type:
Bottle
Pk:
25 g
Seq:
3308
:
:
146.09 g/mol
:
Ambient
:
1993
:
3,III
:
GHS02
:
GHS07
:
60598-17-6
:
C₄H₆F₄O
:
MFCD01075284