Order Entry
United Kingdom
Orders LinkContactUsLinkComponent
3'-Fluoro-4'-hydroxyacetophenone

3'-Fluoro-4'-hydroxyacetophenone

Bottle, 1 g
Catalog # APOSPC0584-1G
Irritant
Supplier:   Apollo Scientific
3'-Fluoro-4'-hydroxyacetophenone
Catalog # APOSPC0584-1G
Supplier:  Apollo Scientific
CAS Number:  403-14-5
3'-Fluoro-4'-hydroxyacetophenone

Bottle, 1 g

CAS Number:  403-14-5
Price
£16.50
Log in for availability
1 * 1 g
Product Details & Documents
Synonyms:  1-(3-Fluoro-4-hydroxyphenyl)-1-ethanone
  • CAS Number:403-14-5
  • Formula:C₈H₇FO₂
  • MDL Number:MFCD00215840
  • Molecular Weight:154.14 g/mol
  • Melting Pt:127…129 °C
  • Storage Temperature:Ambient
Specifications

Specifications

  • Pack type:
    Bottle
  • Pk:
    1 g

Specifications