
3-Fluoro-2-(trifluoromethyl)benzoyl chloride 97%
Supplier: Apollo Scientific
Certificates
About this item
CAS Number:261951-82-0
:
Formula:C₈H₃ClF₄O
MDL Number:MFCD01631458
:226.56 g/mol
Storage Temperature:Ambient
UN:1760
ADR:8,III
:GHS05
Frequently Bought Together