6-Chloro-2-fluoro-3-methylanisole 98%
Supplier: Apollo Scientific
Certificates
About this item
CAS Number:261762-80-5
:
Formula:C₈H₈ClFO
MDL Number:MFCD01631377
:174.60 g/mol
Boiling Pt:66 °C
Storage Temperature:Ambient
Frequently Bought Together