Order Entry
United Kingdom
ContactUsLinkComponent
2-Fluoro-5-methylanisole 98%
Irritant
2-Fluoro-5-methylanisole 98%
Catalog # APOSPC0083-1G
Supplier:  Apollo Scientific
CAS Number:  
Irritant
2-Fluoro-5-methylanisole 98%
Catalog # APOSPC0083-1G
Supplier:  Apollo Scientific
CAS Number:  

Specifications

  • Pack type:
    Bottle
  • Pk:
    1 g
  • Seq:
    3047

Specifications

About this item

 :
 :140.16 g/mol
Boiling Pt:93…95 °C/30mmHg
Density:1.07 g/cm³ (25 °C)
Flash Pt:78 °C
 :GHS07
Storage Temperature:Ambient
CAS Number:63762-78-7
Formula:C₈H₉FO
MDL Number:MFCD00665791