α-Bromo-3-chloro-4-fluorotoluene 97%
Supplier: Apollo Scientific
Certificates
About this item
CAS Number:192702-01-5
:
Formula:C₇H₅BrClF
MDL Number:MFCD01631551
:223.47 g/mol
Boiling Pt:63…65 °C/0.6mmHg
UN:3265
ADR:8,II
:GHS05
:GHS07
Storage Temperature:Ambient
Frequently Bought Together