Methyl quinoline-2-carboxylate 97%
Catalog # APOSOR959530-5G
Supplier: Apollo Scientific
CAS Number:
Specifications
- Pack type:Bottle
- Pk:5 g
- Seq:3173
Specifications
About this item
:
:187.2 g/mol
Storage Temperature:Ambient
CAS Number:19575-07-6
Formula:C₁₁H₉NO₂
MDL Number:MFCD00160630