1-Cyclopropyl-1-phenylmethanamine 97%
Supplier: Apollo Scientific
Certificates
About this item
CAS Number:23459-38-3
:MFCD08445589
Formula:C₁₀H₁₃N
MDL Number:MFCD05191945
:147.22 g/mol
:GHS07
Storage Temperature:Ambient
Frequently Bought Together