Order Entry
United Kingdom
ContactUsLinkComponent
Methyl-3-oxo-4-phenylbutyrate (mixture of isomers) 98%
Methyl-3-oxo-4-phenylbutyrate (mixture of isomers) 98%
Catalog # APOSOR951019-5G
Supplier:  Apollo Scientific
CAS Number:  
Methyl-3-oxo-4-phenylbutyrate (mixture of isomers) 98%
Catalog # APOSOR951019-5G
Supplier:  Apollo Scientific
CAS Number:  

Specifications

  • Pack type:
    Bottle
  • Pk:
    5 g
  • Seq:
    3173
  • Environmentally Preferable:

Specifications

About this item

Synonyms: Methyl 3-oxo-4-phenylbutanoate, Methyl 4-Phenylacetoacetate, 3-Oxo-4-phenylbutyric Acid Methyl Ester, 4-Phenylacetoacetic Acid Methyl Ester
 :
 :192.21 g/mol
Storage Temperature:Ambient
CAS Number:37779-49-0
Formula:C₁₁H₁₂O₃
MDL Number:MFCD03424746