4-Allyloxybenzoic acid 95%
Catalog # APOSOR941983-1G
Supplier: Apollo Scientific
CAS Number:
Specifications
- Pack type:Bottle
- Pk:1 g
- Seq:3047
Specifications
About this item
:
:178.19 g/mol
Storage Temperature:Ambient
CAS Number:27914-60-9
Formula:C₁₀H₁₀O₃
MDL Number:MFCD00526542