Methyl quinoline-5-carboxylate 98%
Catalog # APOSOR940019-1G
Supplier: Apollo Scientific
CAS Number:
Specifications
- Pack type:Bottle
- Pk:1 g
- Seq:3047
Specifications
About this item
:
:187.2 g/mol
Storage Temperature:Ambient
CAS Number:16675-62-0
Formula:C₁₁H₉NO₂
MDL Number:MFCD06204270