Order Entry
United Kingdom
ContactUsLinkComponent
6-Methyl-4,5,6,7-tetrahydro-1,3-benzothiazol-2-amine 95%
6-Methyl-4,5,6,7-tetrahydro-1,3-benzothiazol-2-amine 95%
Catalog # APOSOR939449-1G
Supplier:  Apollo Scientific
CAS Number:  
6-Methyl-4,5,6,7-tetrahydro-1,3-benzothiazol-2-amine 95%
Catalog # APOSOR939449-1G
Supplier:  Apollo Scientific
CAS Number:  

Specifications

  • Pack type:
    Bottle
  • Pk:
    1 g
  • Seq:
    3047

Specifications

About this item

 :
 :168.26 g/mol
Storage Temperature:Ambient
CAS Number:7496-50-6
Formula:C₈H₁₂N₂S
MDL Number:MFCD00805140