Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
£0.00
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
United Kingdom
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
[(1-Ethylpiperidin-4-yl)methyl]amine
[(1-Ethylpiperidin-4-yl)methyl]amine 95%
[(1-Ethylpiperidin-4-yl)methyl]amine 95%
APOSOR939382-1G
:
Apollo Scientific
:
[(1-Ethylpiperidin-4-yl)methyl]amine 95%
APOSOR939382-1G
:
Apollo Scientific
:
Pack type:
Bottle
Pk:
1 g
Seq:
3047
:
:
142.25 g/mol
:
Ambient
:
21168-71-8
:
C₈H₁₈N₂
:
MFCD05022431