Order Entry
United Kingdom
ContactUsLinkComponent
Diethyl(isopropyl)silanol 95%
Diethyl(isopropyl)silanol 95%
Catalog # APOSOR936239-5G
Supplier:  Apollo Scientific
CAS Number:  
Diethyl(isopropyl)silanol 95%
Catalog # APOSOR936239-5G
Supplier:  Apollo Scientific
CAS Number:  

Specifications

  • Pack type:
    Bottle
  • Pk:
    5 g
  • Seq:
    3173

Specifications

About this item

 :
 :146.31 g/mol
Storage Temperature:Ambient
CAS Number:1158830-31-9
Formula:C₇H₁₈OSi
MDL Number:MFCD03093625