Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
£0.00
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
United Kingdom
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
2-Norbornene-2-boronic acid
2-Norbornene-2-boronic acid 95%
2-Norbornene-2-boronic acid 95%
2-Norbornene-2-boronic acid 95%
APOSOR9349-250MG
:
Apollo Scientific
:
2-Norbornene-2-boronic acid 95%
APOSOR9349-250MG
:
Apollo Scientific
:
Pack type:
Bottle
Pk:
250 mg
Seq:
2590
:
:
137.97 g/mol
:
124…126 °C
:
GHS07
:
Ambient
:
871333-98-1
:
C₇H₁₁BO₂
:
MFCD07783853