
4-Hydroxy-3-nitrobenzaldehyde 98%
:3011-34-5
:
:C₇H₅NO₄
:MFCD00007117
:167.12 g/mol
:140…144 °C
:221-141-6
:GHS07
:Ambient
Frequently Bought Together
Acetone ≥99.5%, GPR RECTAPUR®
: VWR Chemicals
Water, GPR RECTAPUR®
: VWR Chemicals
4-Methyl-2-pentanone ≥98%, GPR RECTAPUR®
: VWR Chemicals
Ethanol 96% vol Ph. Eur.
: VWR Chemicals
Ethanol denatured 70% ≥70% (v/v), TechniSolv®
: VWR Chemicals
Dimethyl sulphoxide ≥99.0%, GPR RECTAPUR®
: VWR Chemicals
Acetone ≥99.7%, PESTINORM® for pesticide residue analysis
: VWR Chemicals
Cyclohexane ≥99%, GPR RECTAPUR®
: VWR Chemicals
Silver(I) oxide ≥99% (metals basis)
: Thermo Fisher Scientific
2-Propanol ≥99.9%, HiPerSolv CHROMANORM® for LC-MS
: VWR Chemicals