Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
£0.00
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
United Kingdom
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
3-Methyl-1H-pyrazole-5-carbaldehyde
3-Methyl-1H-pyrazole-5-carbaldehyde 95%
3-Methyl-1H-pyrazole-5-carbaldehyde 95%
3-Methyl-1H-pyrazole-5-carbaldehyde 95%
APOSOR925890-1G
:
Apollo Scientific
:
3-Methyl-1H-pyrazole-5-carbaldehyde 95%
APOSOR925890-1G
:
Apollo Scientific
:
Pack type:
Bottle
Pk:
1 g
Seq:
3047
:
:
110.12 g/mol
:
Ambient
:
3273-44-7
:
MFCD09055186
:
C₅H₆N₂O
:
MFCD00159629