Order Entry
United Kingdom
ContactUsLinkComponent
 
Monomethyl-5-norbornene-2,3-dicarboxylate 95%
  APOSOR924889-5G
 :  
Monomethyl-5-norbornene-2,3-dicarboxylate 95%
  APOSOR924889-5G
 :  

 

  • Pack type:
    Bottle
  • Pk:
    5 g
  • Seq:
    3173

 

 

 :
 :196.20 g/mol
 :Ambient
 :36897-94-6
 :C₁₀H₁₂O₄
 :MFCD00167743