Order Entry
United Kingdom
ContactUsLinkComponent
(±)-2-Bromopropionic acid 98%
Corrosive
Irritant
(±)-2-Bromopropionic acid 98%
Catalog # APOSOR923008-1KG
Supplier:  Apollo Scientific
CAS Number:  
Corrosive
Irritant
(±)-2-Bromopropionic acid 98%
Catalog # APOSOR923008-1KG
Supplier:  Apollo Scientific
CAS Number:  

Specifications

  • Pack type:
    Bottle
  • Pk:
    1 kg
  • Seq:
    4024

Specifications

About this item

 :
 :152.98 g/mol
Storage Temperature:Refrigerator
UN:3265
ADR:8,III
 :GHS05
 :GHS07
CAS Number:598-72-1
Formula:H₃CBrCHCOOH
MDL Number:MFCD00004211