Order Entry
United Kingdom
ContactUsLinkComponent
Diethyl carbonate 98%
Diethyl carbonate 98%
Catalog # APOSOR922483-500G
Supplier:  Apollo Scientific
CAS Number:  
Diethyl carbonate 98%
Catalog # APOSOR922483-500G
Supplier:  Apollo Scientific
CAS Number:  

Specifications

  • Pack type:
    Bottle
  • Pk:
    500 g
  • Seq:
    3645

Specifications

About this item

 :
 :118.13 g/mol
Boiling Pt:126…128 °C
Melting Pt:–43 °C
Density:0.975 g/cm³ (25 °C)
Flash Pt:25 °C
Storage Temperature:Ambient
UN:2366
ADR:3,III
CAS Number:105-58-8
Formula:C₅H₁₀O₃
MDL Number:MFCD00009107