Order Entry
United Kingdom
ContactUsLinkComponent
 
3-Phenylpentan-3-amine hydrochloride 95%
Catalog # APOSOR913327-1G
Supplier:  Apollo Scientific
CAS Number:  
3-Phenylpentan-3-amine hydrochloride 95%
Catalog # APOSOR913327-1G
Supplier:  Apollo Scientific
CAS Number:  

Specifications

  • Pack type:
    Bottle
  • Pk:
    1 g
  • Seq:
    3047

Specifications

About this item

 :
Storage Temperature:Ambient
CAS Number:104177-96-0
Formula:C11H18ClN