Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
£0.00
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
United Kingdom
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
Diethyl phenylphosphonite
Diethyl phenylphosphonite 95%
Diethyl phenylphosphonite 95%
APOSOR911833-25G
:
Apollo Scientific
:
Diethyl phenylphosphonite 95%
APOSOR911833-25G
:
Apollo Scientific
:
Pack type:
Bottle
Pk:
25 g
Seq:
3308
:
:
198.2 g/mol
:
Ambient
:
1638-86-4
:
C₁₀H₁₅O₂P
:
MFCD00009086