Methyl-3-amino-6-chloropyrazine-2-carboxylate 95%
Catalog # APOSOR905907-5G
Supplier: Apollo Scientific
CAS Number:
Specifications
- Pack type:Bottle
- Pk:5 g
- Seq:3173
Specifications
About this item
:
:187.58 g/mol
Storage Temperature:Ambient
CAS Number:1458-03-3
Formula:C₆H₆ClN₃O₂
MDL Number:MFCD00834965