
Isoquinoline-5-sulphonyl chloride hydrochloride 95%
Supplier: Apollo Scientific
Certificates
About this item
CAS Number:105627-79-0
:
Formula:C₉H₇Cl₂NO₂S
MDL Number:MFCD08272853
:264.13 g/mol
Storage Temperature:Refrigerator
UN:3261
ADR:8,III
:GHS05
Frequently Bought Together