Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
£0.00
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
United Kingdom
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
Br-Mmc (=4-bromomethyl-7-methoxycoumarin)
Br-Mmc (=4-bromomethyl-7-methoxycoumarin)
Br-Mmc (=4-bromomethyl-7-methoxycoumarin)
APOSOR8540T-5G
:
Apollo Scientific
:
Br-Mmc (=4-bromomethyl-7-methoxycoumarin)
APOSOR8540T-5G
:
Apollo Scientific
:
Pack type:
Bottle
Pk:
5 g
Seq:
3173
:
:
269.09 g/mol
:
214…217 °C
:
Ambient
:
35231-44-8
:
C₁₁H₉BrO₃
:
MFCD00006869