Benzophenone
Supplier: Apollo Scientific
Jump to:
About this item
Synonyms: Diphenylmethanone, Diphenyl ketone
- CAS Number:119-61-9
- Formula:C₁₃H₁₀O
- MDL Number:MFCD00003076
- Molecular Weight:182.22 g/mol
- Boiling Pt:305 °C
- Melting Pt:47…51 °C
- Flash Pt:138 °C
- Storage Temperature:Ambient
- EINECS:204-337-6
- UN:3077
- ADR:9+EHS,III
2 Options Available Below
Product Details & Documents
Synonyms: Diphenylmethanone, Diphenyl ketone
- CAS Number:119-61-9
- Formula:C₁₃H₁₀O
- MDL Number:MFCD00003076
- Molecular Weight:182.22 g/mol
- Boiling Pt:305 °C
- Melting Pt:47…51 °C
- Flash Pt:138 °C
- Storage Temperature:Ambient
- EINECS:204-337-6
- UN:3077
- ADR:9+EHS,III



