4-[3-(Diphenylmethyl)-1,2,4-oxadiazol-5-yl]piperidine 95+%
Supplier: Apollo Scientific
Certificates
About this item
CAS Number:849925-02-6
:
Formula:C₂₀H₂₁N₃O
MDL Number:MFCD03426081
:319.41 g/mol
Melting Pt:64…67 °C
Storage Temperature:Ambient
Frequently Bought Together