4'-Methoxybiphenyl-3-carbaldehyde
Supplier: Apollo Scientific
Jump to:
About this item
Synonyms: 4'-Methoxy-[1,1'-biphenyl]-3-carboxaldehyde
- CAS Number:118350-17-7
- Formula:C₁₄H₁₂O₂
- MDL Number:MFCD03424627
- Molecular Weight:212.25 g/mol
- Melting Pt:50…53 °C
- Storage Temperature:Ambient
2 Options Available Below
Product Details & Documents
Synonyms: 4'-Methoxy-[1,1'-biphenyl]-3-carboxaldehyde
- CAS Number:118350-17-7
- Formula:C₁₄H₁₂O₂
- MDL Number:MFCD03424627
- Molecular Weight:212.25 g/mol
- Melting Pt:50…53 °C
- Storage Temperature:Ambient



