
Methyl-2-hydroxyisonicotinate 97%
Supplier: Apollo Scientific
Certificates
About this item
CAS Number:89937-77-9
:
Formula:C₇H₇NO₃
MDL Number:MFCD09258780
:153.1400 g/mol
Melting Pt:210…214 °C
:GHS07
Storage Temperature:Ambient
Frequently Bought Together