3-(2,3-Dihydro-5-oxo-(1H,5H)-benzo[ij]quinolizin-6-yl)-(2E)-propenoic acid 95+%
Supplier: Apollo Scientific
Certificates
About this item
CAS Number:386715-44-2
:
Formula:C₁₅H₁₃NO₃
MDL Number:MFCD01567779
:255.28 g/mol
Melting Pt:>255 °C (dec.)
Storage Temperature:Ambient
Frequently Bought Together