2,4-Dichloropyridine
Supplier: Apollo Scientific
Certificates
About this item
CAS Number:26452-80-2
:
Formula:C₅H₃Cl₂N
MDL Number:MFCD00955618
Boiling Pt:189…190 °C
Melting Pt:–1 °C
Density:1.37 g/cm³ (25 °C)
Flash Pt:89 °C
:147.99 g/mol
EINECS:247-717-7
:GHS07
Storage Temperature:Ambient
Frequently Bought Together