Order Entry
United Kingdom
Orders LinkContactUsLinkComponent
 

3,4-Diaminopyridine

Toxic
New Product
Supplier:   Apollo Scientific
3,4-Diaminopyridine
Supplier:  Apollo Scientific
New Product
Certificates
3,4-Diaminopyridine

4 Options Available Below

Product Details & Documents
Synonyms:  3,4-Pyridinediamine, Amifampridine, 3,4-DAP, Pyridine-3,4-diyldiamine, Pyridine-3,4-diamine
  • CAS Number:54-96-6
  • Formula:C₅H₇N₃
  • MDL Number:MFCD00006401
  • Molecular Weight:109.13 g/mol
  • Melting Pt:216…220 °C
  • Storage Temperature:Ambient
  • EINECS:200-220-9
  • UN:2811
  • ADR:6.1,II
Product Family Options

Product Information

  • Pack type
    Pk
    Availability
    Price
  • Bottle
    Log in for availability
    Log in for availability
    1 * 1 g
    1 * 1 g - £16.50
    £16.50
    1 * 1 g
       £16.50

    Specifications:
    New Product
    New Product
  • Bottle
    Log in for availability
    Log in for availability
    1 * 5 g
    1 * 5 g - £18.70
    £18.70
    1 * 5 g
       £18.70

    Specifications:
    New Product
    New Product
  • Bottle
    100 g
    Log in for availability
    Log in for availability
    1 * 100 g
    1 * 100 g - £171.00
    £171.00
    1 * 100 g
       £171.00

  • Bottle
    Log in for availability
    Log in for availability
    1 * 500 g
    1 * 500 g - £843.00
    £843.00
    1 * 500 g
       £843.00

    Specifications:
    New Product
    New Product
Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.