Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
£0.00
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
United Kingdom
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
6-Bromo-2-methyl-3H-imidazo[4,5-b]pyridine
6-Bromo-2-methyl-3H-imidazo[4,5-b]pyridine
6-Bromo-2-methyl-3H-imidazo[4,5-b]pyridine
APOSOR61524-250MG
:
Apollo Scientific
:
6-Bromo-2-methyl-3H-imidazo[4,5-b]pyridine
APOSOR61524-250MG
:
Apollo Scientific
:
Pack type:
Bottle
Pk:
250 mg
Seq:
2590
:
:
212.05 g/mol
:
GHS07
:
Ambient
:
42869-47-6
:
C₇H₆BrN₃
:
MFCD00763284