Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
£0.00
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
United Kingdom
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
1-Benzhydrylazetidin-3-amine
1-Benzhydrylazetidin-3-amine 97%
1-Benzhydrylazetidin-3-amine 97%
1-Benzhydrylazetidin-3-amine 97%
APOSOR6150-1G
:
Apollo Scientific
:
1-Benzhydrylazetidin-3-amine 97%
APOSOR6150-1G
:
Apollo Scientific
:
Pack type:
Bottle
Pk:
1 g
Seq:
3047
:
:
238.34 g/mol
:
GHS07
:
Refrigerator
:
40432-52-8
:
C₁₆H₁₈N₂
:
MFCD03093386