
2-Amino-3-chloropyrazine 97%
Supplier: Apollo Scientific
Certificates
About this item
CAS Number:6863-73-6
:6663-73-6
Formula:C₄H₄ClN₃
MDL Number:MFCD04114305
:129.55 g/mol
Boiling Pt:248 °C
Melting Pt:167…171 °C
:GHS07
Storage Temperature:Ambient
Frequently Bought Together