
1,5,6,7-Tetrahydro-4H-indol-4-one
Catalog # APOSOR61260-25G
Supplier: Apollo Scientific
CAS Number:
Specifications
- Pack type:Bottle
- Pk:25 g
- Seq:3308
Specifications
About this item
:
:135.17 g/mol
Melting Pt:190 °C
:GHS07
Storage Temperature:Refrigerator
CAS Number:13754-86-4
Formula:C₈H₉NO
MDL Number:MFCD00075438