
4-Bromo-1H-pyrazole-5-carbaldehyde
Supplier: Apollo Scientific
Certificates
About this item
CAS Number:287917-97-9
:
Formula:C₄H₃BrN₂O
MDL Number:MFCD00831607
:174.99 g/mol
Melting Pt:178…180 °C
:GHS07
Storage Temperature:Ambient
Frequently Bought Together