Order Entry
United Kingdom
ContactUsLinkComponent
3,4-Dimethylbenzylamine 95%
Corrosive
3,4-Dimethylbenzylamine 95%
Catalog # APOSOR60034-5G
Supplier:  Apollo Scientific
CAS Number:  
Corrosive
3,4-Dimethylbenzylamine 95%
Catalog # APOSOR60034-5G
Supplier:  Apollo Scientific
CAS Number:  

Specifications

  • Pack type:
    Bottle
  • Pk:
    5 g
  • Seq:
    3173

Specifications

About this item

 :
 :135.21 g/mol
Storage Temperature:Ambient
UN:2735
ADR:8,III
 :GHS05
CAS Number:102-48-7
Formula:C₉H₁₃N
MDL Number:MFCD00025577