2-Bromo-5-chlorophenol 97%
Bottle, 5 g
Catalog # APOSOR59899-5G
Supplier: Apollo Scientific
2-Bromo-5-chlorophenol 97%
Catalog # APOSOR59899-5G
Supplier: Apollo Scientific
CAS Number: 13659-23-9
Jump to:
Product Details & Documents
- CAS Number:13659-23-9
- Formula:C₆H₄BrClO
- MDL Number:MFCD00142870
- Molecular Weight:207.45 g/mol
- Melting Pt:63…65 °C
- Storage Temperature:Ambient
Specifications
Specifications
- Pack type:Bottle
- Pk:5 g



