Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
£0.00
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
United Kingdom
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
2-Chloro-6-(methylamino)pyrazine
2-Chloro-6-(methylamino)pyrazine
2-Chloro-6-(methylamino)pyrazine
APOSOR59340-500MG
:
Apollo Scientific
:
2-Chloro-6-(methylamino)pyrazine
APOSOR59340-500MG
:
Apollo Scientific
:
Pack type:
Bottle
Pk:
500 mg
Seq:
2725
:
:
80…83 °C
:
143.5700 g/mol
:
GHS07
:
Ambient
:
848366-38-1
:
C₅H₆ClN₃
:
MFCD09878383