
3-Cyano-4-methoxy-2-pyridone 98+%
Supplier: Apollo Scientific
Certificates
About this item
CAS Number:21642-98-8
:MFCD00975442
Formula:C₇H₆N₂O₂
MDL Number:MFCD08059559
:150.14 g/mol
Melting Pt:275…276 °C
:GHS07
Storage Temperature:Ambient
Frequently Bought Together