

2-Ethoxybenzoyl chloride 98%
Supplier: Apollo Scientific
Certificates
About this item
CAS Number:42926-52-3
:
Formula:C₉H₉ClO₂
MDL Number:MFCD00016348
:184.62 g/mol
Boiling Pt:114…118 °C/5mmHg
Density:1.19 g/cm³ (25 °C)
Flash Pt:>110 °C
Storage Temperature:Ambient
EINECS:256-003-4
UN:3265
ADR:8,II
:GHS05
:GHS07
Frequently Bought Together