L(-)-Malic acid 99%
Supplier: Apollo Scientific
Certificates
About this item
CAS Number:97-67-6
:
Formula:C₄H₆O₅
MDL Number:MFCD00064213
:134.09 g/mol
Melting Pt:101…103 °C
EINECS:202-601-5
:GHS07
Storage Temperature:Ambient
Frequently Bought Together