Order Entry
United Kingdom
ContactUsLinkComponent
 
3-Cyanobenzenesulphonamide 97%
Catalog # APOSOR480670-5G
Supplier:  Apollo Scientific
CAS Number:  
3-Cyanobenzenesulphonamide 97%
Catalog # APOSOR480670-5G
Supplier:  Apollo Scientific
CAS Number:  

Specifications

  • Pack type:
    Bottle
  • Pk:
    5 g
  • Seq:
    3173

Specifications

About this item

 :
 :182.2 g/mol
Storage Temperature:Ambient
CAS Number:3118-68-1
Formula:C₇H₆N₂O₂S
MDL Number:MFCD02089633