Diethyl terephthalate
Catalog # APOSOR480605-100G
Supplier: Apollo Scientific
CAS Number:
Specifications
- Pack type:Bottle
- Pk:100 g
- Seq:3401
Specifications
About this item
:
:222.24 g/mol
Storage Temperature:Ambient
CAS Number:636-09-9
Formula:C₁₂H₁₄O₄
MDL Number:MFCD00039891