2-(3-Pyrrolidinyl)acetic acid hydrochloride
Catalog # APOSOR47890-1G
Supplier: Apollo Scientific
CAS Number:
Specifications
- Pack type:Bottle
- Pk:1 g
- Seq:3047
Specifications
About this item
:
:165.62 g/mol
Storage Temperature:Ambient
CAS Number:89203-64-5
Formula:C₆H₁₂ClNO₂
MDL Number:MFCD06795694