C-[1-(4-Chlorophenyl)cyclopentyl]methylamine
Catalog # APOSOR470260-5G
Supplier: Apollo Scientific
CAS Number:
Specifications
- Pack type:Bottle
- Pk:5 g
- Seq:3173
Specifications
About this item
:
:209.72 g/mol
Storage Temperature:Ambient
CAS Number:75180-51-7
Formula:C₁₂H₁₆ClN
MDL Number:MFCD07801114