Order Entry
United Kingdom
Orders LinkContactUsLinkComponent
 

2-Iodophenol 98%

Irritant
Supplier:   Apollo Scientific
2-Iodophenol 98%
Supplier:  Apollo Scientific
Certificates
2-Iodophenol 98%

3 Options Available Below

Product Details & Documents
  • CAS Number:533-58-4
  • Formula:C₆H₅IO
  • MDL Number:MFCD00013963
  • Molecular Weight:220.01 g/mol
  • Boiling Pt:186…187 °C/160mmHg
  • Melting Pt:37…43 °C
  • Density:1.947 g/cm³ (25 °C)
  • Flash Pt:>110 °C
  • Storage Temperature:Refrigerator
  • EINECS:208-569-9
Product Family Options

Product Information

  • Pack type
    Pk
    Availability
    Price
  • Bottle
    25 g
    Log in for availability
    Log in for availability
    1 * 25 g
    1 * 25 g - £19.80
    £19.80
    1 * 25 g
       £19.80

  • Bottle
    100 g
    Log in for availability
    Log in for availability
    1 * 100 g
    1 * 100 g - £49.50
    £49.50
    1 * 100 g
       £49.50

  • Bottle
    500 g
    Log in for availability
    Log in for availability
    1 * 500 g
    1 * 500 g - £244.00
    £244.00
    1 * 500 g
       £244.00