Order Entry
United Kingdom
ContactUsLinkComponent
Diethyl azodicarboxylate 40% in toluene
Diethyl azodicarboxylate 40% in toluene
Catalog # APOSOR46008-100G
Supplier:  Apollo Scientific
CAS Number:  
Diethyl azodicarboxylate 40% in toluene
Catalog # APOSOR46008-100G
Supplier:  Apollo Scientific
CAS Number:  

Specifications

  • Pack type:
    Bottle
  • Pk:
    100 g
  • Seq:
    3401

Specifications

About this item

 :
 :174.16 g/mol
Density:0.96 g/cm³ (25 °C)
Flash Pt:41 °C
Storage Temperature:Refrigerator
UN:1993
ADR:3,II
 :GHS02
 :GHS07
 :GHS08
CAS Number:1972-28-7
Formula:C₆H₁₀N₂O₄
MDL Number:MFCD00009103