4-Hydroxy-4-phenylpiperidine 98+%
Supplier: Apollo Scientific
Certificates
About this item
CAS Number:40807-61-2
:
Formula:C₁₁H₁₅NO
MDL Number:MFCD00006000
:177.25 g/mol
Melting Pt:157…161 °C
EINECS:255-089-0
:GHS07
Storage Temperature:Ambient
Frequently Bought Together